3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 98%
Supplier: Thermo Fisher Scientific
Warning
436550050 436550250 436551000
ACRO436550050EA 178 EUR
ACRO436550050 ACRO436550250 ACRO436551000
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 98%
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Formula: C₃₂H₅₈N₂O₇S
Melting Pt: 267.2…271.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00012116
CAS Number: 75621-03-3
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure