3-Amino-1-Boc-piperidine 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate
436600010 436600050
ACRO436600010EA 190 EUR
ACRO436600010 ACRO436600050
3-Amino-1-Boc-piperidine 97%
3-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861219
CAS Number: 144243-24-3
Order Now
Specification Test Results
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %