2-Chloropyrimidine-5-carboxyaldehyde 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-Chloropyrimidine-5-carboxaldehyde, 2-Chloropyrimidine-5-carbaldehyde
438450010 438450050
ACRO438450010EA 185 EUR
ACRO438450010 ACRO438450050
2-Chloropyrimidine-5-carboxyaldehyde 97%
2-Chloropyrimidine-5-carboxyaldehyde
Formula: C₅H₃ClN₂O
MW: 142.55 g/mol
Melting Pt: 113…118 °C
Storage Temperature: Ambient
MDL Number: MFCD10696891
CAS Number: 933702-55-7
Order Now
Specification Test Results
Appearance White to yellow Powder
Infrared spectrum Conforms
GC ≥96.0 %