4,5-Dimethoxy-2-nitrobenzyl bromide 97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1-(Bromomethyl)-2-nitro-4,5-dimethoxybenzene, 1-(Bromomethyl)-4,5-dimethoxy-2-nitrobenzene, 6-Nitroveratryl bromide
440770010 440770050
ACRO440770010EA 94 EUR
ACRO440770010 ACRO440770050
4,5-Dimethoxy-2-nitrobenzyl bromide 97%
4,5-Dimethoxy-2-nitrobenzyl bromide
Formula: C₉H₁₀BrNO₄
MW: 276.086 g/mol
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00192064
CAS Number: 53413-67-5
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance Yellow to brown Crystalline powder or needles
Infrared spectrum Conforms
HPLC ≥96.0 %