4-(Tetrazol-5-yl)phenylboronic acid 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: [4-(1H-Tetrazol-5-yl)phenyl]boronic acid, 4-(2H-1,2,3,4-TETRAZOL-5-YL)PHENYLBORONIC ACID, (4-(2H-Tetrazol-5-yl)phenyl)boronic acid, Boronic acid, B-[4-(1H-tetrazol-5-yl)phenyl]-
445570010 445570050
ACRO445570010EA 123 EUR
ACRO445570010 ACRO445570050
4-(Tetrazol-5-yl)phenylboronic acid 97%
4-(Tetrazol-5-yl)phenylboronic acid
Formula: C₇H₇BN₄O₂
MW: 189.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739099
CAS Number: 179942-55-3
Order Now
Specification Test Results
Appearance White to off-white Solid
Infrared spectrum Conforms
HPLC ≥95.0 %
NMR (1H-NMR) Consistent with structure