2-Iodosobenzoic acid 98%
Supplier: Thermo Fisher Scientific
Warning
446030010 446030050
ACRO446030010EA 45.3 EUR
ACRO446030010 ACRO446030050
2-Iodosobenzoic acid 98%
2-Iodosobenzoic acid
Formula: C₇H₅IO₃
Melting Pt: 200…204 °C
Storage Temperature: Freezer
MDL Number: MFCD00002401
CAS Number: 304-91-6
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Iodometric 97.5 to 102.5 %
HPLC ≥97.5 %