Sodium chlorate 99% ACS
Supplier: Thermo Fisher Scientific
Danger
446411000 446415000
ACRO446411000EA 30.8 EUR
ACRO446411000 ACRO446415000
Sodium chlorate 99% ACS
Sodium chlorate
Formula: ClNaO₃
MW: 106.44 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 248…261 °C
Density: 2.49 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003479
CAS Number: 7775-09-9
UN: 1495
ADR: 5.1 II
Merck Index: 13,08670
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Titration ≥99.0 %
Chloride (Cl) ≤0.005 %
Bromate (BrO3) ≤0.015 %
Sulfate (SO4) ≤0.001 %
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.002 %
Potassium (K) ≤0.01 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.005 %