Benzonitrile for HPLC
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Cyanobenzene, Phenylcyanide
446910010 446911000
ACRO446910010EA 234 EUR
ACRO446910010 ACRO446911000
Benzonitrile for HPLC
Benzonitrile
Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 188…191 °C (760 mmHg)
Melting Pt: –13 °C
Density: 1.01 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00001770
CAS Number: 100-47-0
UN: 2224
ADR: 6.1 II
Merck Index: 13,01098
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.03 %
GC ≥99.0 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %