Lithium-tert-butanolate 8% (w/w) 0.9 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
446841000 446848000
446841000.EA 72.4 EUR
446841000. 446848000.
Lithium-tert-butanolate 8% (w/w) 0.9 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Lithium tert-butanolate
Formula: C₄H₉LiO
MW: 80.06 g/mol
Density: 0.86 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00050479
CAS Number: 1907-33-1
UN: 2924
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear yellow to brown Solution (May develop some turbidity or precipitate)
Strength 0.80 to 1.10 M