Mercury(I) chloride ACS
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Dimercury dichloride, Mercurous chloride, Calomel
447591000 447595000
ACRO447591000EA 79.5 EUR
ACRO447591000 ACRO447595000
Mercury(I) chloride ACS
Mercury (I) chloride
Formula: Cl₂Hg₂
MW: 472.09 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 400 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011043
CAS Number: 10112-91-1
UN: 2025
ADR: 6.1 III
Merck Index: 12,05951
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Titration Iodometric ≥99.5 %
Sulfate (SO4) ≤0.01 %
Mercuric Chloride ≤0.01 % (HgCl2)
Residue after reduction ≤0.02 %