Atenolol 98%
Supplier: Thermo Fisher Scientific
Synonyms: 2-[4-(2-Hydroxy-3-isopropylaminopropoxy)phenyl]acetamide
449550010 449550050 449550250
ACRO449550010EA 63.3 EUR
ACRO449550010 ACRO449550050 ACRO449550250
Atenolol 98%
Atenolol
Formula: C₁₄H₂₂N₂O₃
MW: 266.34 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00057645
CAS Number: 29122-68-7
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 151 to 155 °C
Titration with HClO4 ≥97.5 %