Sodium hydroxide, micropearls, extra pure
Supplier: Thermo Fisher Scientific
Danger
451130050 451130250
ACRO451130050EA 50.6 EUR
ACRO451130050 ACRO451130250
Sodium hydroxide, micropearls, extra pure
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 318 °C
Density: 2,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Appearance White Pearls
Particle size 20 to 40 mesh
Titration with HCl ≤2.5 % (Na2CO3)