Tetrabutylammonium fluoride 75% in water
Supplier: Thermo Fisher Scientific
Danger
451800250 451801000 451805000
ACRO451800250EA 48.3 EUR
ACRO451800250 ACRO451801000 ACRO451805000
Tetrabutylammonium fluoride 75% in water
Tetrabutylammonium fluoride
Formula: C₁₆H₃₆FN
MW: 261.47 g/mol
Density: 0.953 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011747
CAS Number: 429-41-4
UN: 2922
ADR: 8 II
Order Now
Specification Test Results
Appearance Colorless to light yellow Viscous liquid
Water 20 to 30 %
Titration with HClO4 70 to 80 %