2-Hydrazinopyrimidine 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-Hydrazinylpyrimidine
453050010 453050050
ACRO453050010EA 71 EUR
ACRO453050010 ACRO453050050
2-Hydrazinopyrimidine 98%
2-Hydrazinopyrimidine
Formula: C₄H₆N₄
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01249337
CAS Number: 7504-94-1
Order Now
Specification Test Results
Appearance White to off-white to light brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %