Potassium nitrite 97%, extra pure
Supplier: Thermo Fisher Scientific
Danger
453190010 453190050
ACRO453190010EA 86.3 EUR
ACRO453190010 ACRO453190050
Potassium nitrite 97%, extra pure
Potassium nitrite
Formula: KNO₂
MW: 85.1 g/mol
Melting Pt: 387 °C
Storage Temperature: Ambient
MDL Number: MFCD00011408
CAS Number: 7758-09-0
UN: 1488
ADR: 5.1 II
Merck Index: 13,07734
Order Now
Specification Test Results
Appearance White to light yellow Powder or crystals
Titration excess KMnO4 96.0 to 104.0 %
Arsenic (As) ≤1 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 % (10 % in water)