Test Kits, photometric, potassium, LCK cuvette tests
Supplier: Hach
LCK328 LCK228
705-3064EA 126 EUR
705-3064 HACHLCK228
Test Kits, photometric, potassium, LCK cuvette tests
Environmental Test Kits
Cuvette tests for water testing applications. The cuvette is a closed system which is safe for the user and the environment.
  • Precise and reliable measurement values
  • Maximum safety for users, due to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C (protect from light)
Storage conditions: 15 to 25 °C (protect from light)
Accessories information: Cuvette tests are officially approved for many legally required consent limits and with the help of Addista standard solutions and round-robin test solutions, they provide the assurance to be needed.