Palmatine chloride hydrate, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

361615-100MG 361615-500MG
SIAL361615-100MGEA 64 EUR
SIAL361615-100MG SIAL361615-500MG
Palmatine chloride hydrate, Sigma-Aldrich®
Palmatine chloride hydrate
Formula: C₂₁H₂₂CINO₄·H₂O
CAS Number: 171869-95-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR