General purpose gloves, Senti-Flex, A360
Supplier: Portwest
A360B8RS A360B8RM A360B8RXXL A360B8RL A360B8RXL
112-4594EA 25.7 EUR
112-4594 112-4595 112-4598 112-4596 112-4597
General purpose gloves, Senti-Flex, A360
Gloves
These ultra-thin nylon gloves with PU coating on palm and fingertips are ideal for precision handling.
  • PU dipped for abrasion resistance
  • Ultimate dexterity and breathability
Certifications: EN 388:2016 (3121X)