Ball valve lockouts, Prinzing
Supplier: Brady
800110 800111
118-0534EA 49.21 EUR
118-0534 118-0535
Ball valve lockouts, Prinzing
Security Locks
These devices lock out pipes from 50 to 200 mm in either open/closed (small) or in closed (large) position.
  • Two halves of the device encompass the ball valve lever to secure it from inadvertent activation of the valve
  • Made of ultra-tough PP
Max. shackle diameter: 7,24 mm
Min. shackle clearance: 38,00 mm
Pipe diameter: 14,50 - 52,50 mm (small); 50 to 200 mm (large).