Miniature circuit breaker lockouts
Supplier: Brady
090854 090853 090845 805853 090851 090844 090850 090847 090848
BRDY090854EA 117.94 EUR
BRDY090854 BRDY090853 BRDY090845 118-0538 BRDY090851 BRDY090844 BRDY090850 BRDY090847 BRDY090848
Miniature circuit breaker lockouts
Security Locks
These miniature circuit breakers are a safe and effective method for locking out European circuit breakers as they fit most of the existing types. Recommended for use in combination with a padlock for added safety.
  • Miniature breaker lockout unique in its kind: Requires no tools to install
  • Lockout easily installs with the aid of a push button
  • Tie-bar style lockout has a convenient thumb wheel for quick application
  • Suitable for single and multi-pole breakers
  • Made from glass-filled nylon
  • One padlock can be applied
Max. shackle Ø: 7 mm