Leuco crystal violet, powder
Supplier: MP Biomedicals
Warning
Synonyms: 4,4',4''-Methanetriyltris(N,N-dimethylaniline), Tris(4-dimethylaminophenyl)methane, 4,4',4''-Methylidynetris(N,N-dimethylaniline)
0215519505 0215519525
ICNA0215519505EA 62 EUR
ICNA0215519505 ICNA0215519525
Leuco crystal violet, powder
Leuco crystal violet
Soluble in benzene (0,5/20 - clear).
  • λmax 260 nm(lit.)
Caution: Research Use Only (RUO).
Formula: C₂₅H₃₁N₃
MW: 373.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008314
CAS Number: 603-48-5