Analytical HPLC columns, Ultra silica (USP L3)
Supplier: Restek
9100311 9100312 9100315 9100331 9100332 9100335 9100351 9100352 9100355 9100361 9100362 9100365 9100511 9100512 9100515 9100521 9100522 9100525 9100531 9100532 9100535 9100551 9100552 9100555 9100561 9100562 9100565 9100571 9100572 9100575 910050210 910050212 9100312-700 9100313-700 9100315-700 9100332-700 9100333-700 9100335-700 9100352-700 9100353-700 9100355-700 9100362-700 9100363-700 9100365-700 9100512-700 9100513-700 9100515-700 9100522-700 9100523-700 9100525-700 9100532-700 9100533-700 9100535-700 9100552-700 9100553-700 9100555-700 9100562-700 9100563-700 9100565-700 9100572-700 9100573-700 9100575-700 910031E 910033E 910035E 910036E 910051E 910052E 910053E 910055E 910056E 910057E
REST9100311EA 0 EUR
REST9100311 REST9100312 REST9100315 REST9100331 REST9100332 REST9100335 REST9100351 REST9100352 REST9100355 REST9100361 REST9100362 REST9100365 REST9100511 REST9100512 REST9100515 REST9100521 REST9100522 REST9100525 REST9100531 REST9100532 REST9100535 REST9100551 REST9100552 REST9100555 REST9100561 REST9100562 REST9100565 REST9100571 REST9100572 REST9100575 REST910050210 REST910050212 REST9100312-700 REST9100313-700 REST9100315-700 REST9100332-700 REST9100333-700 REST9100335-700 REST9100352-700 REST9100353-700 REST9100355-700 REST9100362-700 REST9100363-700 REST9100365-700 REST9100512-700 REST9100513-700 REST9100515-700 REST9100522-700 REST9100523-700 REST9100525-700 REST9100532-700 REST9100533-700 REST9100535-700 REST9100552-700 REST9100553-700 REST9100555-700 REST9100562-700 REST9100563-700 REST9100565-700 REST9100572-700 REST9100573-700 REST9100575-700 REST910031E REST910033E REST910035E REST910036E REST910051E REST910052E REST910053E REST910055E REST910056E REST910057E
Analytical HPLC columns, Ultra silica (USP L3)
Chromatography Columns
Ultra silica columns made of high-purity, type-B silica are designed for selective and reliable HPLC applications.
  • Base-deactivated spherical silica
  • Minimises activity and creates high-density bonding