Capillary columns, MXT®-1701
Supplier: Restek
72020 72023 72040 72051 72052 72053 72055 72066 72070 72082 72085 72088
REST72020EA 0 EUR
REST72020 REST72023 REST72040 REST72051 REST72052 REST72053 REST72055 REST72066 REST72070 REST72082 REST72085 REST72088
Capillary columns, MXT®-1701
Chromatography Columns
MXT®-1701 Siltek®-treated stainless steel columns are general purpose columns for alcohols, oxygenates, PCB congeners (e.g., Aroclor mixes), and pesticides.
  • Midpolarity Crossbond® phase
  • Temperature range -20 to 280 °C
  • Equivalent to USP G46 phase
Coil diameter: 114,3 mm