Anti-GFAP Mouse Monoclonal Antibody (CF770) [clone: GA-5]
Supplier: Biotium
BNC700167-500 BNC700167-100
BNC700167-500EA 560 EUR
BNC700167-500 BNC700167-100
Anti-GFAP Mouse Monoclonal Antibody (CF770) [clone: GA-5]
Antibodies
This MAb recognizes a protein of ~50 kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins. GFAP is specifically found in astroglia. GFAP is a very popular marker for localizing benign astrocyte and neoplastic cells of glial origin in the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS.
Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: GA-5
Conjugation: CF770
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Cow, Human, Mouse, Pig, Rabbit, Rat