Anti-ICAM3 Mouse Monoclonal Antibody (CF488A) [clone: 186-2G9]

Supplier: Biotium
BNC880178-500 BNC880178-100
BNC880178-500EA 560 EUR
BNC880178-500 BNC880178-100
Anti-ICAM3 Mouse Monoclonal Antibody (CF488A) [clone: 186-2G9]
Antibodies
Recognizes the D1 domain of an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3 (Leucocyte Workshop VI). CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb is excellent for staining of formalin/paraffin tissues.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: 186-2G9
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ICAM3
Antigen name CD50
Environmentally Preferable
Antigen synonyms ICAMR|Intercellular adhesion molecule 3 (ICAM3)
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 488A
Clone Clone 186-2G9
Reactivity Human
Host Mouse
Gene ID 354563
Isotype IgG2b kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR