Anti-MVP Mouse Monoclonal Antibody (CF647) [clone: 1014]
Supplier: Biotium
BNC470224-500 BNC470224-100
BNC470224-500EA 560 EUR
BNC470224-500 BNC470224-100
Anti-MVP Mouse Monoclonal Antibody (CF647) [clone: 1014]
Antibodies
Recognizes a protein of 104 kDa-110 kDa, characterized as major vault protein (MVP). Vaults are large ribonucleoprotein particles (RNPs) present in all eukaryotic cells. They have a complex morphology, including several small molecules of RNA, but a single protein species. The MVP accounts for >70% of their mass. Their shape is reminiscent of the nucleopore central plug. Treatment of cells with estradiol increases the amount of MVP in nuclear extract. The hormone-dependent interaction of vaults with ER is prevented in vitro by sodium molybdate. Antibodies to estrogen, progesterone andMore Product Information
Order Now
SPECIFICATIONS
Antigen symbol MVP
Antigen name Major Vault Protein
Environmentally Preferable
Antigen synonyms MVP|VAULT1|Major Vault Protein|Lung resistance-related protein (LRP)
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 647
Clone Clone 1014
Reactivity Human
Host Mouse
Gene ID 632177
Isotype IgG1 kappa
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide