Anti-MVP Mouse Monoclonal Antibody (CF640R) [clone: 1032]
Supplier: Biotium
BNC400225-500 BNC400225-100
BNC400225-500EA 560 EUR
BNC400225-500 BNC400225-100
Anti-MVP Mouse Monoclonal Antibody (CF640R) [clone: 1032]
Antibodies
Recognizes a protein of 104 kDa-110 kDa, characterized as major vault protein (MVP). Vaults are large ribonucleoprotein particles (RNPs) present in all eukaryotic cells. They have a complex morphology, including several small molecules of RNA, but a single protein species. The MVP accounts for >70% of their mass. Their shape is reminiscent of the nucleopore central plug. Treatment of cells with estradiol increases the amount of MVP in nuclear extract. The hormone-dependent interaction of vaults with ER is prevented in vitro by sodium molybdate. Antibodies to estrogen, progesterone andMore Product Information