Anti-MYB Mouse Monoclonal Antibody (CF594) [clone: MYB286]
Supplier: Biotium
BNC940286-500 BNC940286-100
BNC940286-500EA 560 EUR
BNC940286-500 BNC940286-100
Anti-MYB Mouse Monoclonal Antibody (CF594) [clone: MYB286]
Antibodies
The highly leukemogenic avian retrovirus E26 contains two oncogenes, v-Myb and v-Ets, which are expressed together as a fusion protein. The cellular homolog of v-Myb, designated c-Myb, encodes a transcription factor. Deletion or disruption of a negative regulatory domain mapping within the carboxy-terminal domain of c-Myb results in enhanced transactivating capacity and in parallel, leads to activation of its ability to transform hemopoietic cells. c-Myb is expressed preferentially, but not exclusively, in immature hemopoietic cells and its expression decreases as cells differentiate.