Anti-AFP Mouse Monoclonal Antibody (CF647) [clone: C3]
Supplier: Biotium
BNC470354-500 BNC470354-100
BNC470354-500EA 560 EUR
BNC470354-500 BNC470354-100
Anti-AFP Mouse Monoclonal Antibody (CF647) [clone: C3]
Antibodies
This antibody recognizes an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as alpha-fetoprotein (AFP) (ISOBM TD-2 workshop). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C3
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Dog, Human, Monkey, Pig