Anti-KRT8 Mouse Monoclonal Antibody (Biotin) [clone: K8/383]

Supplier: Biotium
BNCB0383-500 BNCB0383-100
BNCB0383-500EA 560 EUR
BNCB0383-500 BNCB0383-100
Anti-KRT8 Mouse Monoclonal Antibody (Biotin) [clone: K8/383]
Antibodies
Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). CK8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin 8 and 18. CK8 exists on several types of normal and neoplastic epithelia, including many ductal and glandular epithelia such as colon, stomach, small intestine, trachea, and esophagus as well as in transitional epithelium. Anti-CK8 does not react with skeletal muscle or nerve cells. Epithelioid sarcoma, chordoma, and adamantinoma show strong positivity corresponding to that of simple epithelia (with antibodies against CK8, CK18 and CK19). Reportedly, anti-CK8 is useful for the differentiation of lobular ring-like, perinuclear from ductal peripheral-predominant carcinoma of the breast.

Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: K8/383
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR