Anti-MUC1 Mouse Monoclonal Antibody (CF405S) [clone: E29]
Supplier: Biotium
BNC040478-500 BNC040478-100
BNC040478-500EA 560 EUR
BNC040478-500 BNC040478-100
Anti-MUC1 Mouse Monoclonal Antibody (CF405S) [clone: E29]
Antibodies
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinomas. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver.
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: E29
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human