Anti-ESR1 Mouse Monoclonal Antibody (Purified) [clone: ER505]

Supplier: Biotium
BNUB0505-500 BNUB0505-100
BTIUBNUB0505-500EA 511 EUR
BTIUBNUB0505-500 BTIUBNUB0505-100
Anti-ESR1 Mouse Monoclonal Antibody (Purified) [clone: ER505]
Antibodies
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. This MAb is excellent for staining of formalin-fixed, paraffin-embedded breast carcinomas.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ER505
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR