Anti-ESR1 Mouse Monoclonal Antibody (CF647) [clone: ER505]

Supplier: Biotium
BNC470505-500 BNC470505-100
BTIUBNC470505-500EA 560 EUR
BTIUBNC470505-500 BTIUBNC470505-100
Anti-ESR1 Mouse Monoclonal Antibody (CF647) [clone: ER505]
Antibodies
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. This MAb is excellent for staining of formalin-fixed, paraffin-embedded breast carcinomas.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ER505
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR