Anti-MUC1 Mouse Monoclonal Antibody (CF640R) [clone: MUC1/520]

Supplier: Biotium
BNC400520-500 BNC400520-100
BNC400520-500EA 560 EUR
BNC400520-500 BNC400520-100
Anti-MUC1 Mouse Monoclonal Antibody (CF640R) [clone: MUC1/520]
Antibodies
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinomas. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: MUC1/520
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol MUC1
Environmentally Preferable
Antigen name Mucin 1 / EMA / Episialin / CD227
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 640R
Clone Clone MUC1/520
Reactivity Human
Host Mouse
Gene ID 89603
Isotype IgG2a kappa
ImmunoChemistry Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR