Anti-VWF Mouse Monoclonal Antibody (CF488A) [clone: 3E2D10 VWF635]
Supplier: Biotium
BNC880646-500 BNC880646-100
BNC880646-500EA 560 EUR
BNC880646-500 BNC880646-100
Anti-VWF Mouse Monoclonal Antibody (CF488A) [clone: 3E2D10 VWF635]
Antibodies
von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascularMore Product Information
Order Now
SPECIFICATIONS
Antigen symbol VWF VWF
Environmentally Preferable
Antigen name von Willebrand Factor von Willebrand Factor
Antigen synonyms F8VWF|von Willebrand Disease (vWD)|Factor VIII Related Antigen|Coagulation Factor VIII|von Willebrand Antigen 2 F8VWF|von Willebrand Disease (vWD)|Factor VIII Related Antigen|Coagulation Factor VIII|von Willebrand Antigen 2
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Conjugation CF® 488A CF® 488A
Clone Clone 3E2D10 VWF635 Clone 3E2D10 VWF635
Reactivity Human Human
Host Mouse Mouse
Gene ID 440848 440848
Isotype IgGs IgGs
Western blot Yes Yes
ImmunoChemistry Yes Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide