Anti-Il6 (Mouse) Rat Monoclonal Antibody (CF405S) [clone: MP5-20F3]
Supplier: Biotium
BNC040663-500 BNC040663-100
BNC040663-500EA 560 EUR
BNC040663-500 BNC040663-100
Anti-Il6 (Mouse) Rat Monoclonal Antibody (CF405S) [clone: MP5-20F3]
Antibodies
IL-6 is a potent lymphoid cell growth factor that stimulates the growth and survivability of certain B-cells and T-cells. IL-6 plays a role in host defense, acute phase reactions, immune responses, and hematopoiesis. IL-6 is expressed by T-cells, B-cells, monocytes, fibroblasts, hepatocytes, endothelial cells and keratinocytes.
Type: Primary
Antigen: Il6 (Mouse)
Clonality: Monoclonal
Clone: Clone MP5-20F3
Conjugation: CF405S
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse