Anti-NCAM1 Mouse Monoclonal Antibody [clone: 123C3.D5 123A8]

Supplier: Biotium
BNUB0693-500 BNUB0693-100
BNUB0693-500EA 511 EUR
BNUB0693-500 BNUB0693-100
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 123C3.D5 123A8]
Antibodies
This MAb reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Three isoforms of neural cell adhesion molecule (NCAM) are produced by differential splicing of the RNA transcript from a single gene. The 135 kDa isoform is the basic molecule, which is glycosylated or sialylated to produce the mature species. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas). It is also expressed on some mesodermally derived tumors (rhabdomyosarcoma). Anti-CD56 plays an important role in the diagnosis of nodal and nasal NK/T-cell lymphomas.

  • Purified

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 123C3.D5 123A8
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR