Anti-NCAM1 Mouse Monoclonal Antibody (CF640R) [clone: 123C3.D5 123A8]
Supplier: Biotium
BNC400693-500 BNC400693-100
BNC400693-500EA 560 EUR
BNC400693-500 BNC400693-100
Anti-NCAM1 Mouse Monoclonal Antibody (CF640R) [clone: 123C3.D5 123A8]
Antibodies
This MAb reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Three isoforms of neural cell adhesion molecule (NCAM) are produced by differential splicing of the RNA transcript from a single gene. The 135 kDa isoform is the basic molecule, which is glycosylated or sialylated to produce the mature species. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas). It is also expressed on some mesodermally derived tumors (rhabdomyosarcoma). Anti-CD56 plays an important role in the diagnosis of nodal and nasal NK/T-cell lymphomas.
Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 123C3.D5 123A8
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human