Anti-IGL Mouse Monoclonal Antibody (CF594) [clone: LcN-2 ICO-106]

Supplier: Biotium
BNC940735-500 BNC940735-100
BNC940735-500EA 560 EUR
BNC940735-500 BNC940735-100
Anti-IGL Mouse Monoclonal Antibody (CF594) [clone: LcN-2 ICO-106]
Antibodies
This MAb is specific to lambda light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the lambda light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoidMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol IGL
Environmentally Preferable
Antigen name Lambda Light Chain
Antigen synonyms Paraprotein|BJP|Mcg Marker|Bence Jones Protein|IGLC 1/2/3
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 594
Clone Clone LcN-2 ICO-106
Reactivity Human
Host Mouse
Gene ID 449585
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR