Anti-PRLR Mouse Monoclonal Antibody (CF568) [clone: B6.2 PRLR742]

Supplier: Biotium
BNC680744-500 BNC680744-100
BNC680744-500EA 560 EUR
BNC680744-500 BNC680744-100
Anti-PRLR Mouse Monoclonal Antibody (CF568) [clone: B6.2 PRLR742]
Antibodies
This antibody recognizes a protein of 70 kDa, identified as prolactin receptor. Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor), which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription.

Type: Primary
Antigen:More Product Information
Order Now

SPECIFICATIONS

Antigen symbol PRLR PRLR
Antigen name Prolactin Receptor Prolactin Receptor
Environmentally Preferable
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Conjugation CF® 568 CF® 568
Clone Clone B6.2 PRLR742 Clone B6.2 PRLR742
Reactivity Human Human
Host Mouse Mouse
Gene ID 368587 368587
Isotype IgGs IgGs
ImmunoChemistry Yes Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR