Anti-BCL2L1 Mouse Monoclonal Antibody (Biotin) [clone: 2H12]

Supplier: Biotium
BNCB0751-500 BNCB0751-100
BNCB0751-500EA 560 EUR
BNCB0751-500 BNCB0751-100
Anti-BCL2L1 Mouse Monoclonal Antibody (Biotin) [clone: 2H12]
Antibodies
Recognizes a protein of 27 kDa, identified as the Bcl-X protein. This MAb shows no cross-reaction with Bcl-2 or Bax protein. Bcl-X has two isoforms, Bcl-XL (long), a 241 amino acid protein which suppresses cell death. And Bcl-XS (short), a 178 amino acid protein lacking a 63 amino acid domain which functions as a dominant inhibitor of Bcl-2. This MAb reacts with both Bcl-XS and Bcl-XL proteins.

Type: Primary
Antigen: BCL2L1
Clonality: Monoclonal
Clone: 2H12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR