Anti-HSPD1 Mouse Monoclonal Antibody (CF640R) [clone: HSPD1/780]

Supplier: Biotium
BNC400780-500 BNC400780-100
BNC400780-500EA 560 EUR
BNC400780-500 BNC400780-100
Anti-HSPD1 Mouse Monoclonal Antibody (CF640R) [clone: HSPD1/780]
Antibodies
Recognizes a 60 kDa protein, identified as the heat shock protein 60 (hsp60). A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp), more appropriately called as stress response proteins (srp). Hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial hsp60, but also against its mammalian homolog.

Order Now

SPECIFICATIONS

Antigen symbol HSPD1
Antigen name HSP60
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 640R
Clone Clone HSPD1/780
Reactivity Hamster,Pig,Rabbit,Monkey,Sheep,Chicken,Human,Bovine,Dog,Rat,Mouse
Host Mouse
Gene ID 595053
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR