Anti-TYRP1 Mouse Monoclonal Antibody (CF640R) [clone: TA99 TYRP1/807]
Supplier: Biotium
BNC400808-500 BNC400808-100
BNC400808-500EA 560 EUR
BNC400808-500 BNC400808-100
Anti-TYRP1 Mouse Monoclonal Antibody (CF640R) [clone: TA99 TYRP1/807]
Antibodies
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.
Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: TA99 TYRP1/807
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Mouse