Anti-WT1 Mouse Monoclonal Antibody (CF405S) [clone: WT1/857]

Supplier: Biotium
BNC040857-500 BNC040857-100
BNC040857-500EA 560 EUR
BNC040857-500 BNC040857-100
Anti-WT1 Mouse Monoclonal Antibody (CF405S) [clone: WT1/857]
Antibodies
Recognizes a 47-55 kDa-tumor suppressor protein, identified as Wilm's Tumor (WT1) protein. The antibody reacts with all isoforms of the full-length WT1 and also identifies WT1 lacking exon 2-encoded amino acids, frequently found in subsets of sporadic Wilm s tumors.WT1, a sporadic and familial pediatric kidney tumor, is genetically heterogeneous. Wilm s tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelium, andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol WT1
Environmentally Preferable
Antigen synonyms WAGR|FWT1|AWT1|NPHS4|WT1|GUD|Wilms'tumor 1
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 405S
Clone Clone WT1/857
Reactivity Human,Rat,Mouse
Host Mouse
Gene ID 591980
Isotype IgGs
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR