Anti-CD44 Mouse Monoclonal Antibody (CF647) [clone: HCAM/918]

Supplier: Biotium
BNC470918-500 BNC470918-100
BNC470918-500EA 560 EUR
BNC470918-500 BNC470918-100
Anti-CD44 Mouse Monoclonal Antibody (CF647) [clone: HCAM/918]
Antibodies
Recognizes a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes (Leucocyte Typing Workshop V). Its epitope is resistant to digestion by trypsin and chymotrypsin. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion and stromal binding. CD44 immunostaining is commonly used for the discrimination of urothelial transitional cell carcinoma in-situ from non-neoplastic changes in the urothelium.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: HCAM/918
Conjugation: CF647
Epitope:
Host: Mouse
Reactivity: Human, Non-human primates
Isotype: IgG2a, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR