Anti-CD31 Rat Monoclonal Antibody [clone: MEC13.3]

Supplier: Novus Biologicals
NB600-1475 NB600-1475SS
NOVUNB600-1475EA 642 EUR
NOVUNB600-1475 NOVUNB600-1475SS
Anti-CD31 Rat Monoclonal Antibody [clone: MEC13.3]
Antibodies
Rat Monoclonal CD31/PECAM1 Antibody (MEC13.3). Tested Applications: Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR