Anti-ICAM3 Mouse Monoclonal Antibody (Purified) [clone: ICAM3/1019]

Supplier: Biotium
BNUB1019-500 BNUB1019-100
BNUB1019-500EA 511 EUR
BNUB1019-500 BNUB1019-100
Anti-ICAM3 Mouse Monoclonal Antibody (Purified) [clone: ICAM3/1019]
Antibodies
Recognizes an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3. CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb is excellent for staining of formalin/paraffin tissues.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: ICAM3/1019
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR