Anti-ICAM3 Mouse Monoclonal Antibody (CF488A) [clone: ICAM3/1019]

Supplier: Biotium
BNC881019-500 BNC881019-100
BNC881019-500EA 560 EUR
BNC881019-500 BNC881019-100
Anti-ICAM3 Mouse Monoclonal Antibody (CF488A) [clone: ICAM3/1019]
Antibodies
Recognizes an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3. CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb is excellent for staining of formalin/paraffin tissues.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: ICAM3/1019
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol ICAM3
Antigen name CD50
Environmentally Preferable
Antigen synonyms ICAMR|Intercellular adhesion molecule 3 (ICAM3)
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 488A
Clone Clone ICAM3/1019
Reactivity Human
Host Mouse
Gene ID 354563
Isotype IgG2b kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR