Anti-TFRC Mouse Monoclonal Antibody (Biotin) [clone: TFRC/1149]
Supplier: Biotium
BNCB1149-500 BNCB1149-100
BNCB1149-500EA 560 EUR
BNCB1149-500 BNCB1149-100
Anti-TFRC Mouse Monoclonal Antibody (Biotin) [clone: TFRC/1149]
Antibodies
This antibody recognizes a ~90-95 kDa protein which is identified as cell surface transferrin receptor (CD71), a disulfide-bonded homodimeric glycoprotein of 180-190 kDa. This MAb is highly specific to CD71 and shows no cross-reaction with other related proteins. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. CD71/Transferrin receptor has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinicalMore Product Information