Anti-KRT19 Mouse Monoclonal Antibody (CF647) [clone: KRT19/799 KRT19/800]
Supplier: Biotium
BNC471150-500 BNC471150-100
BNC471150-500EA 560 EUR
BNC471150-500 BNC471150-100
Anti-KRT19 Mouse Monoclonal Antibody (CF647) [clone: KRT19/799 KRT19/800]
Antibodies
This antibody recognizes a protein of 40 kDa, identified as cytokeratin-19 (CK19), which is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK1More Product Information